Please login to your online account to display your discounted pricing
Thermo Scientific™ Crystal Plates and Spare Parts for the Thermo Scientific™ iD7 ATR Accessory
Choose the crystal plate to meet specific application and budget requirements
$8,772.79 - $8,772.79
Products
Catalog Number Mfr. No. Product Type Price Quantity    
14-387-801
thermo scientific™
869168800
iD7 AR Diamond Plate, AR-coated Each for $8,772.79
14-387-802
thermo scientific™
869168900
iD7 XR Diamond Plate, uncoated Each for $8,772.79
Description & Specifications
Configure the iD7 ATR accessory with one or more of the crystal plates:
  • Diamond (AR-coated or uncoated) for liquids and all solids
  • ZnSe for liquids and soft solids
  • Germanium for highly absorbing materials like black rubbers
Spare part kit includes iD7 ATR tip and volatiles cover.