Please login to your online account to display your discounted pricing
Thermo Scientific™ Counterstains
$481.87 - $481.87
Products
Catalog Number Mfr. No. Quantity Type Price Quantity    
14-390-24
thermo scientific™
1931492
16 oz. (475mL) Eosin Y Counterstain Case of 4 Each for $481.87
Description & Specifications
  • EA-50: Polychromatic cytoplasmic stain that differentiates cellular components with pink, green, and blue-green hues
  • EA-65: Polychromatic cytoplasmic stain with lower concentration of Light Green that allows better expression of the eosin-stained components; ideal for thicker, non-gynecological preparations
  • Eosin Y: Fast-acting, ready-to-use cytoplasmic counterstain that colors tissues bright pink for easy identification
  • Orange G-6 (OG-6): Detects keratin in cytoplasm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.