missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Clear Seal Diamond Heat Sealing Film

Clear polymer heat seal is ideal for fluorescence and colorimetric applications

Supplier:  Thermo Scientific™ AB3799

 View more versions of this product

Catalog No. AB-3799



Description

Description

Clear polymer heat seal is ideal for fluorescence and colorimetric applications

  • Suitable for PCR in screw-down or clip-down heated lid thermal cyclers
  • Good optical clarity, ideal for QPCR and other fluorescence applications
  • Peelable
  • Some solvent resistance
  • Seal integrity range: -80°C to +120°C

Specifications

Specifications

Clear
Clear Seal Diamond - Clear polymer material; peelable seal; excellent optical clarity; some solvent resistance
Non-sterile
96
370 m x 78 mm
Roll
Heat Seal
Polypropylene
No
All well shapes
Life Science Reagent Resistant
Polyethylene
Not Autoclavable
1 Roll
Storage, PCR, qPCR, Molecular Diagnostics
ALPS 5000 & ALPS 3000
Abgene
No
-80°C to +120°C
Polypropylene, Polyethylene, Cyclic Olefin Copolymer, Polystyrene
SDS
Documents

Documents

Promotions

Promotions