Please login to your online account to display your discounted pricing
Thermo Scientific™ CL10 Trunnion Rotors
Choice of two systems for use with Thermo Scientific™ CL10 centrifuge
$1,506.28 - $1,506.28
Products
Catalog Number Mfr. No. Price Quantity    
05-376-224
thermo scientific™
11210015
Pack for $1,506.28
Description & Specifications
  • 16 × 15mL rotor with unsealed carriers (4 trunnion rings with 4 places per ring)
  • 8 × 15mL rotor with sealed carriers (4 trunnion rings with 2 places per ring)
  • Rotor head only also available
  • Max. speed/RCF: 4000rpm/2575 x g
Spare parts available online. For rotor 05-376-224: trunnions (set of 4) 05-376-225 and unsealed carriers (set of 8).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.