Please login to your online account to display your discounted pricing
Thermo Scientific™ CL10 and Primo™ Compact Clinical Centrifuge Packages
Benchtop centrifuges optimized for typical applications with rotor and adapters
$6,840.34 - $6,840.34
Specifications
Refrigerated No
Voltage 120V
Rotor 4x100mL swinging bucket rotor
Rotor Included Yes
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
75-489-998
thermo scientific™
75489998
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
75-849-998
thermo scientific™
75849998
Each for $6,840.34
Description & Specifications
Specifications
Refrigerated No
Voltage 120V
Rotor 4x100mL swinging bucket rotor
Rotor Included Yes
Packaged for immediate use in the clinical laboratory
Blood analysis and separation; urine analysis; separation of other body fluids; DNA/RNA separations; PCR work-up
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.