missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Citadel™ Tissue Processor Accessories

For use with the Thermo Scientific Citadel Tissue Processor

$7,333.81 - $7,333.81

Specifications

Quantity Each
View More Specs

Products 1
Catalog Number Mfr. No. For Use With Product Type Price Quantity  
Catalog Number Mfr. No. For Use With Product Type Price Quantity  
69810053
View Documents
Thermo Scientific™
69810053
Tissue processor Wax Bath Assembly Each for $7,333.81
 
Specifications

Specifications

Each