Fisherbrand™ Castaloy™ Two-Prong Extension Clamps
Flat, vinyl- or fiberglass-covered jaws
Manufacturer: Fisherbrand™ 00176Q
Includes: Vinyl and fiberglass sleeves, extension arm O.D.: 0.43 in. (1.1cm)
Description
Flat, vinyl- or fiberglass-covered jaws
Hold beakers and flasks firmly at any angle, variable distance from support.
- One jaw is fixed, other opens against a stainless-steel compression spring
- Opening is gradual to maximum angle, without binding
- Noncorrosive
- Easy-to-clean clamps are nickel-plated zinc with seamless brass extension tube, brass compressing screw, and pivot pin
- Without clamp holder
Castaloy clamps and holders should not be subjected to direct flame or other intense heat sources. At continuous temperatures in excess of 150°C (302°F), some significant distortion can occur. Fiberglass sleeves are recommended for all applications above 100°C (212°F).
Specifications
Fiberglass and Vinyl sleeve | |
22.86 cm | |
6.35 cm |
9 in. | |
2.5 in. | |
Extension Clamp |