Fisherbrand™ Castaloy™ Three-Prong Extension Clamps
Hold irregular shapes at any angle, surely and safely
Manufacturer: Fisherbrand™ 00191Q
Description
Sturdy extension clamps allow you to hold objects with irregular shapes at any angle, safely and securely.
- Two separate adjustment screws ensure even, precise pressure on glass and minimize chance of breakage
- Clamps are easily adjustable to vary holding angle, distance from stand
Specifications
Fiberglass and Vinyl sleeve | |
17 cm | |
2.5 cm | |
Extension Clamp |
6.5 in. | |
1 in. | |
Extension arm O.D.: 0.31 in. (0.8cm) |