Fisherbrand™ Castaloy™ Regular or Heavy-Duty Pinchcocks
Complete closure
Manufacturer:  Fisherbrand™ 00210Q
 View more versions of this product
Catalog No. 05849AQ
Description
Description
  • Do not cut tubing, rust, or corrode
05-849AQ
  • Regular pinchcock opens when plunger is compressed
  • Accommodate 0.25 in. (6mm) O.D. heavy-wall tubing
05-849BQ
  • Heavy-duty model works well with stiff tubing
  • Used in gas and water connections
  • Specifications
    Specifications
    To compress a flexible tube to control the flow of fluid through it
    Accommodate 0.25 in. O.D. heavy wall tubing
    Regular Pinchcock