Please login to your online account to display your discounted pricing
Thermo Scientific™ Bluing Reagents
Reagent for Hematoxylin Bluing
$274.23 - $274.23
Specifications
Type Bluing Reagent
For Use With (Application) Blues hematoxylin-stained nuclei
View More Specs
 Alert
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products
Catalog Number Mfr. No. Quantity Price Quantity    
14-390-15
thermo scientific™
1931423
1 gal. Each for $274.23
Description & Specifications
Specifications
Type Bluing Reagent
For Use With (Application) Blues hematoxylin-stained nuclei
  • Blues hematoxylin-stained nuclei
  • Gentle action ensures no specimen loss or lifting of material as can happen with Scott's tap water
  • Completely ready-to-use and thoroughly pH-controlled for effective bluing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.