Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ Phenyl HPLC Columns


Alternative selectivity of peptides and proteins

Manufacturer: thermo scientific™  72405151030

 View more versions of this product

Catalog No. 03-051-739

  • / Each
 

Description & Specifications

Specifications

Particle Size 5μm
Length (Metric) 150mm
Stationary Phase Phenyl
Diameter (Metric) Inner 1mm