Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ Phenyl HPLC Columns


Alternative selectivity of peptides and proteins

Manufacturer: thermo scientific™  72405254630

 View more versions of this product

Catalog No. 03-051-743

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 250mm
Particle Size 5μm
Stationary Phase Phenyl
Diameter (Metric) Inner 4.6mm