Thermo Scientific™ Barnstead™ ROpure LP™ Membranes
$3,046.29 - $3,046.29
Specifications
Feedwater Temperature (English) 77°F
Feedwater Temperature (Metric) 25°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Feedwater Temperature (English) Feedwater Temperature (Metric) Flow Rate Price Quantity  
Catalog Number Mfr. No. Feedwater Temperature (English) Feedwater Temperature (Metric) Flow Rate Price Quantity  
09-050-37 Thermo Scientific™
D6318
77°F 25°C 0.18 Each for $3,046.29
Specifications
Description
  • Flowrates in table are for new membranes under these conditions:
      25°C (77°F) feedwater temperature
    • Feedwater composition of 1000ppm NaCl at a pH of 6.0 to 6.5
    • Stated design specifications for operating pressure and feedwater recovery
  • Membrane life expectancy will vary with feedwater conditions from one to three years
†Can also be used with discontinued Ultrapure™ systems. Flowrates: 16 gal./hr. (60L/hr.) for 09-034-177B membrane; 29 gal./hr. (110L/hr.) for 09-034-177C; 41 gal./hr. (157L/hr.) for 09-034-177D.
Description & Specifications
Specifications
77°F
25°C