Please login to your online account to display your discounted pricing
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Manufacturer: thermo scientific™  D50231
 View more versions of this product
Catalog No. 09-034-18
  • / Each
Description & Specifications
Specifications
Resin Type Activated Carbon and mixed-bed ion exchange resin
Product Type Organic/ion exchange cartridge
For Use With (Equipment) EasyPure™ II compact ultrapure water system
Media Type Mixed Bed Ion Exchange