Please login to your online account to display your discounted pricing
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Manufacturer: thermo scientific™  D50230
 View more versions of this product
Catalog No. 09-034-17
  • / Each
Description & Specifications
Specifications
Media Type macroreticular
Product Type Organic/colloidal removal
For Use With (Equipment) EasyPure™ II system, 2/3 Macroreticular Resin and 1/3 Granular, Activated Carbon
Resin Type 2/3 Macroreticular resin
Feedwater Source Deionized
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.