Please login to your online account to display your discounted pricing
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
$233.88 - $293.21
Products
Catalog Number Mfr. No. Media Type For Use With (Equipment) Product Type Resin Type Price Quantity    
09-034-17
thermo scientific™
D50230
macroreticular EasyPure™ II system, 2/3 Macroreticular Resin and 1/3 Granular, Activated Carbon Organic/colloidal removal 2/3 Macroreticular resin Each for $240.85
09-034-16
thermo scientific™
D50229
Ultrapure Mixed Bed EasyPure™ II water purification system Organicfree/organic Removal Mixed bed Each for $293.21
09-034-18
thermo scientific™
D50231
Mixed Bed Ion Exchange EasyPure™ II compact ultrapure water system Organic/ion exchange cartridge Activated Carbon and mixed-bed ion exchange resin Each for $233.88
Description & Specifications
  • Easypure™ II Systems require pretreated feedwater
  • From the chart, select appropriate cartridges based on application and feedwater type
Cartridges and filters must be ordered separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.