Please login to your online account to display your discounted pricing

Thermo Scientific™ Barnstead™ Cartridge Kits


Manufacturer: thermo scientific™  D50254

 View more versions of this product

Catalog No. 09-050-248

  • / Each
 

Description & Specifications

Specifications

Product Type Water purification cartridge
Shelf Life 2 Years
For Use With (Equipment) Nanopure Infinity Systems
Model D50254
Includes One 09-050-226 Tap/DI Feed Pretreatment, 09-050-246 Organicfree cartridge, two 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters
Feedwater Source Deionized or Tap
Accessories

2 Years