Please login to your online account to display your discounted pricing
Thermo Scientific™ Barnstead™ Automatic Controls for Electrically Heated Classic Stills
$8,231.96 - $8,231.96
Specifications
Includes All valves, low Water cut off and start/stop control
View More Specs
Products
Catalog Number Mfr. No. Capacity Product Type For Use With (Equipment) Price Quantity    
09-050-55
thermo scientific™
G2100
1 to 2, 3.8 to 7.6 Automatic Control Electrically heated classic still, 24 Hours/day operation Each for $8,231.96
09-050-57
thermo scientific™
G2110
5, 19 Classic Still Automatic control for electrically, storage tanks CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Includes All valves, low Water cut off and start/stop control