Thermo Scientific™ ART™ Non-Filtered Pipet Tips with MicroPoint™ Tip Design
MicroPoint design means the tip profile is nearly 50% smaller than standard blunt-end tips and significantly more narrow than beveled-end tips
$214.74 - $214.74
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Tip Style Packaging Sterility Price Quantity  
Catalog Number Mfr. No. Model Tip Style Packaging Sterility Price Quantity  
21-236-77C Thermo Scientific™
MB3542
XLP Extended Rack Nonsterile Pack of 768 Each for $214.74
Case of 4 Pack for $858.96
Specifications
Description
  • Same superior quality and performance as all Thermo Scientific ART pipet tips
  • Crystal clear tips allow you to easily see transparent samples
  • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting the desired volume of liquid
  • ART 10 tips are ideal for low volume samples
  • The extended length ART 10 REACH™ and ART 1000 REACH prevent contamination of samples, tubes, and pipetters
  • Ultra Micro and Pure Ultra Micro are also well suited for work with low-volume samples
  • ART 200 are equivalent to the “Universal Yellow“ tip and have a reference mark at 10μL
ART Non-Filtered tips are available in yellow and blue colors, graduated, beveled tips and large volumes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.