Thermo Scientific™ SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
$290.00 - $358.10
Specifications
Filtered | Non-Filtered |
---|---|
Color | Natural |
Clean Claims | DNase, RNase, DNA, ATP, and Endotoxin Free |
Compatibility | Rainin™ LTS™ LiteTouch™ |
Format | Hinged Rack |
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity | |||
21-402-553
|
Thermo Scientific™
372105HR |
20L, Non-Filtered, Sterile, Hinged Rack | 20 μL | Red |
Pack of 960 Each for $307.06
Case of 5 Pack for $1,535.30 |
|
21-402-555
|
Thermo Scientific™
372205HR |
20L, Non-Filtered, Non-sterile, Hinged Rack | 20 μL | Red |
Pack of 960 Each for $290.00
Case of 5 Pack for $1,450.00 |
|
21-402-564
|
Thermo Scientific™
375105HR |
200L, Non-Filtered, Sterile, Hinged Rack | 200 μL | Green |
Pack of 960 Each for $305.18
Case of 5 Pack for $1,525.90 |
|
21-402-566
|
Thermo Scientific™
375205HR |
200L, Non-Filtered, Non-sterile, Hinged Rack | 200 μL | Green |
Pack of 960 Each for $290.00
Case of 5 Pack for $1,450.00 |
|
21-402-574
|
Thermo Scientific™
373105HR |
300L, Non-Filtered, Sterile, Hinged Rack | 300 μL | Orange |
Pack of 960 Each for $358.10
Case of 5 Pack for $1,790.50 |
|
21-402-576
|
Thermo Scientific™
373205HR |
300L, Non-Filtered, Non-sterile, Hinged Rack | 300 μL | Orange |
Pack of 960 Each for $338.20
Case of 5 Pack for $1,691.00 |
|
21-402-584
|
Thermo Scientific™
378105HR |
1000L, Non-Filtered, Sterile, Hinged Rack | 1000 μL | Blue |
Pack of 768 Each for $324.93
Case of 4 Pack for $1,299.72 |
|
21-402-586
|
Thermo Scientific™
378205HR |
1000L, Non-Filtered, Non-sterile, Hinged Rack | 1000 μL | Blue |
Pack of 768 Each for $313.90
Case of 4 Pack for $1,255.60 |
|
Description
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
- Low-retention tips maximize sample delivery
- Hinged rack provides easy one-hand access to tips
- Color-coded inserts allows quick identification of tip volume
Specifications
Non-Filtered | |
DNase, RNase, DNA, ATP, and Endotoxin Free | |
Hinged Rack | |
SoftFit L |
Natural | |
Rainin™ LTS™ LiteTouch™ | |
Rainin™ LTS™ LiteTouch™ |