Please login to your online account to display your discounted pricing
Thermo Scientific™ ART™ Barrier SoftFit-L™ Pipette Tips
For use with the Rainin™ LTS™ LiteTouch™ pipettes
$343.74 - $441.29
Specifications
For Use With (Equipment) Rainin™ LTS LiteTouch pipettes
Product Type ART™ Barrier Tip
Sterility Sterile
Tip Style ART Barrier Pipette Tips Filter
View More Specs
Products
Catalog Number Mfr. No. Tip Style Packaging Volume (Metric) Price Quantity    
21-236-90
thermo scientific™
MB2789
ART Barrier Pipette Tips Filter Tray 1200μL Pack of 800 Each for $394.94
Case of 4 Pack for $1,579.76
21-402-162
thermo scientific™
MB2275
ART Barrier Pipette Tips Filter Rack 500μL Pack of 800 Each for $343.74
Case of 4 Pack for $1,374.96
21402488
thermo scientific™
MB274905
ART Barrier Pipette Tips Filter Rack 20μL Pack of 960 Each for $441.29
Case of 5 Pack for $2,206.45
21402489
thermo scientific™
2769-05
ART Barrier Pipette Tips Filter Rack 200μL Pack of 960 Each for $441.29
Case of 5 Pack for $2,206.45
21402490
thermo scientific™
MB277905
ART Barrier Pipette Tips Filter Rack 1200μL Pack of 768 Each for $441.29
Case of 4 Pack for $1,765.16
21-403-00
thermo scientific™
MB2749
ART Barrier Pipette Tips Filter Tray 20μL Pack of 960 Each for $427.48
Case of 5 Pack for $2,137.40
21-403-01
thermo scientific™
MB2769
ART Barrier Pipette Tips Filter Tray 200μL Pack of 960 Each for $394.94
Case of 5 Pack for $1,974.70
Description & Specifications
Specifications
For Use With (Equipment) Rainin™ LTS LiteTouch pipettes
Product Type ART™ Barrier Tip
Sterility Sterile
Tip Style ART Barrier Pipette Tips Filter
The SoftFit-L System is compatible with Rainin LTS pipettes made by Rainin Instrument LCC.
  • Available in low retention
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Manufactured according to the industry's highest quality standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.