Please login to your online account to display your discounted pricing

Thermo Scientific™ ART™ Barrier Pipette Tips with Breakthru Economy Packaging

$230.90 - $406.94

Specifications

For Use With (Equipment) Research grade pipetters
Packaging Rack
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Product Type Tip Style Sterility Volume (Metric) Price Quantity    

21-403-04

 
thermo scientific™
MB2149P50
Tip ART Barrier, MicroPoint Sterile 0.1 to 20μL Pack of 960 Each for $406.94
Case of 10 Pack for $4,069.40

21-403-05

 
thermo scientific™
MB355150
Tip ART Barrier, MicroPoint Sterile 10 to 200μL Pack of 960 Each for $248.65
Case of 10 Pack for $2,486.50

21-403-06

 
thermo scientific™
MB355250
Tip ART Barrier, MicroPoint Nonsterile 10 to 200μL Pack of 960 Each for $230.90
Case of 10 Pack for $2,309.00
Description & Specifications

Specifications

For Use With (Equipment) Research grade pipetters
Packaging Rack
Color Clear

  • The stackable Breakthru™ packaging uses 85% less packaging materials and 53% less space
  • Breakthru packaging is universal to all 9mm trays and works with Art™ self-sealing barrier tips Tips are racked
  • ART 20P and Pure 200 are sterile; ART 200 are nonsterile

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.