Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size


Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples

Manufacturer: thermo scientific™  164899

 View more versions of this product

Catalog No. DX164899

  • / Each
 

Description & Specifications

Specifications

Particle Size 5μm
Type Acclaim PepMap C18 100A nanoViper HPLC Column
Length (Metric) 50mm
Diameter (Metric) Inner 1000μm
Length (English) 2 in.