Please login to your online account to display your discounted pricing
Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples
$1,401.94 - $1,401.94
Products
Catalog Number Mfr. No. Price Quantity    
DX164899
thermo scientific™
164899
Each for $1,401.94
DX164901
thermo scientific™
164901
Each for $1,401.94
DX164900
thermo scientific™
164900
Each for $1,401.94
DX164902
thermo scientific™
164902
Each for $1,401.94
Description & Specifications
  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Inside diameter 300μm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.