Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size


Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples

Manufacturer: thermo scientific™  164901

 View more versions of this product

Catalog No. DX164901

  • / Each
 

Description & Specifications

Specifications

Length (English) 2 in.
Diameter (Metric) Inner 300μm
Type Acclaim PepMap C18 100A nanoViper HPLC Column
Length (Metric) 50mm
Particle Size 5μm