Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size


Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples

Manufacturer: thermo scientific™  164900

 View more versions of this product

Catalog No. DX164900

  • / Each
 

Description & Specifications

Specifications

Type Acclaim PepMap C18 100A nanoViper HPLC Column
Length (English) 6 in.
Particle Size 5μm
Diameter (English) Inside 1000μm
Length (Metric) 150mm