Please login to your online account to display your discounted pricing

Thermo Scientific™ Accessory Rotors for Centra-CL2 Centrifuges Four-Place Swinging Bucket Rotor Packages

$2,612.72 - $2,619.49

Specifications

For Use With (Equipment) Centra CL2 centrifuge
No. of Positions 4.0
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Max. RCF Max. Speed Type Price Quantity    

05-112-112

 
thermo scientific™
523680F
5 to 10mL 2150 3900rpm Rotor Each for $2,612.72

05-112-109

 
thermo scientific™
523670F
15 and 50mL Each for $2,619.49
Description & Specifications

Specifications

For Use With (Equipment) Centra CL2 centrifuge
No. of Positions 4.0

Two packages are available offering a combination of 002360F rotor and Aerocarrier™ tube inserts.

Tissue culture package contains:

  • Rotor and four inserts each for 15 and 50mL Falcon™ and Corning™ conical tubes
Blood tube package consists of:
  • Rotor plus four inserts for 7 to 10mL blood tubes and four inserts for 5 to 7mL blood tubes

Meet OSHA Bloodborne Pathogen Final Rule

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.