Please login to your online account to display your discounted pricing
Thermo Scientific™ Accessory Rotors for Centra-CL2 Centrifuges Four-Place Swinging Bucket Rotor Packages
Manufacturer: thermo scientific™  523670F
 View more versions of this product
Catalog No. 05-112-109
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Centra CL2 centrifuge
Max. Speed 3400rpm
Max. RCF 1950
Type Rotor
No. of Positions 4.0
Capacity (Metric) 15 and 50mL