Thermo Scientific™ Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Choice of accessories for maximum flexibility
Supplier: Thermo Scientific™ 1910083
Description
Choice of accessories for maximum flexibility
UV Light
- Provides safe and easy method for cabinet disinfection
- Factory-installed; order with cabinet
- Included with packages
- Stainless-steel armrests (set of two) provide ergonomic forearm support and extended reach inside cabinet with less airflow disruption; included with packages
- Footrest provides ergonomic foot and posture support during seated use
- Bar and 12 hooks
- For hanging IV bags near interior ceiling of cabinet
- Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
- Method recommended by NSF/ANSI™ 49 for externally exhausting Class II, Type A2 biological safety cabinets
- Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
- Factory-installed; must be ordered with cabinet
Specifications
Interior Tubing Retainer Clips |