Thermo Scientific™ Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Choice of accessories for maximum flexibility
Manufacturer:  Thermo Scientific™ 1910179
 View more versions of this product
Catalog No. 13-261-361
Description
Description
Choice of accessories for maximum flexibility
UV Light
  • Provides safe and easy method for cabinet disinfection
  • Factory-installed; order with cabinet
  • Included with packages
Armrest and Footrest
  • Stainless-steel armrests (set of two) provide ergonomic forearm support and extended reach inside cabinet with less airflow disruption; included with packages
  • Footrest provides ergonomic foot and posture support during seated use
IV Bag Holder Kit
  • Bar and 12 hooks
  • For hanging IV bags near interior ceiling of cabinet
Valves
  • For standard applications or universal piping
  • Standard-application valves have long stem pipe fitting appropriate for side-access media services
  • Short piping stem on universal-piping valves will not penetrate cabinet side wall
  • Combustible gas valve: Rated for combustible gas dispensing inside cabinet chamber
  • Noncombustible gas valve: Rated for noncombustible gas dispensing inside cabinet chamber
  • Vacuum valve: Rated for routing vacuum inside cabinet chamber
Thimble Exhaust Duct Transition
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Method recommended by NSF/ANSI™ 49 for externally exhausting Class II, Type A2 biological safety cabinets
Universal Piping for Exhaust Systems
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Factory-installed; must be ordered with cabinet
Specifications
Specifications
Hanging shelf