Please login to your online account to display your discounted pricing
Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats
Round or square capped mats for sealing 96-well storage plates, available in autoclavable and piercable formats.
Manufacturer: thermo scientific™  AB0427
 View more versions of this product
Catalog No. AB-0427
  • / Pack of 5 Each
Description & Specifications
Specifications
For Use With 96-well PCR plate
No. per Pack 5.0
Material TPE
No. of Wells 96
Autoclavable Yes
Certifications/Compliance Certified free from DNase, RNase and Human Genomic DNA
Temperature Range (English) -4° to 266°F
Temperature Range (Metric) -20° to 130°C
Product Type Sealing Mat