Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats

Round or square capped mats for sealing 96-well storage plates, available in autoclavable and piercable formats.

$181.81 - $402.76

Specifications

Autoclavable Yes
Certifications/Compliance Certified free from DNase, RNase and Human Genomic DNA
Material TPE
No. per Pack 5.0
Product Type Sealing Mat
View More Specs
Products
Catalog Number Mfr. No. For Use With No. of Wells Price Quantity    

AB-0427

 
thermo scientific™
AB0427
96-well PCR plate 96 Pack of 5 Each for $181.81

AB-0770

 
thermo scientific™
AB0770
384-well PCR microplate 384 Pack of 5 Each for $402.76
Description & Specifications

Specifications

Autoclavable Yes
Certifications/Compliance Certified free from DNase, RNase and Human Genomic DNA
Material TPE
No. per Pack 5.0
Product Type Sealing Mat
Temperature Range (English) -4° to 266°F
Temperature Range (Metric) -20° to 130°C

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Round or square capped mats for sealing
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or piercable formats
  • Pierceable version:
    • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
    • Suitable for UV Irradiation and Gamma Irradiation
    • Requires 6N for piercing
    • Temperature Range: -20°C to +55°C
    Autoclavable version:
  • Suitable for UV Irradiation and Gamma Irradiation
  • Temperature Range: -20°C to +130°C
  • Hazardous: No

    Storage Condition: Ambient

    Certified free from DNase, RNase and human genomic DNA.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.