Thermo Scientific™ ABgene™ Thermo-Fast™ 96-Well Semi-Skirted PCR Plates
Patented segmented design allows plate to be cut into 24- and 48-well sections
$189.45 - $365.70
Specifications
Certifications/Compliance Certified free from human DNA, RNases, and DNases
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
AB-0900P Thermo Scientific™
AB0900P
Pack of 25 Each for $228.06
AB-0900Y Thermo Scientific™
AB0900Y
Pack of 25 Each for $228.06
AB-0900W Accessories Thermo Scientific™
AB0900W
Pack of 25 Each for $323.21
AB-0900 Accessories Thermo Scientific™
AB0900
Pack of 25 Each for $246.48
AB-0900G Accessories Thermo Scientific™
AB0900G
Pack of 25 Each for $231.55
AB-0900B Accessories Thermo Scientific™
AB0900B
Pack of 25 Each for $231.55
AB-0990 Thermo Scientific™
AB0990
Pack of 25 Each for $189.45
BC0900W Thermo Scientific™
BC0900W
Pack of 25 Each for $365.70
BC0900 Thermo Scientific™
BC0900
Pack of 25 Each for $353.42
Specifications
Description
  • Virgin polypropylene
  • Raised rim, thin-wall design
  • Clear well bottom for sample visibility
  • Compatible with all major thermal cyclers
  • 300μL maximum well volume (when used with adhesive and heat seals)
  • Cut-away corner and alphanumeric grid
  • Available in a rigid version for robotic handling applications; rigid version cannot be cut
  • Patented segmented plate design allows plates to be cut into 24- and 48-well sections
  • Semi-skirt adds rigidity and allows for labeling or bar coding
  • Cut corner: H1
Certifications
Certified free from DNase, RNase, and human genomic DNA.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Certified free from human DNA, RNases, and DNases