Please login to your online account to display your discounted pricing
Thermo Scientific™ ABgene™ Thermo-Fast™ 96 PCR Detection Plate MKIII
96-well low-profile semi-skirted PCR plate for the fast block of Life Technologies PCR and qPCR thermal cyclers. Supplied with barcodes for sample and plate traceability.
$228.24 - $372.39
Specifications
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
For Use With (Application) PCR, qPCR, RT-PCR
Volume (Metric) Well 100μL
Well Color Clear
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Color Type Price Quantity    
AB1900
thermo scientific™
AB1900
Clear Pack of 25 Each for $228.24
AB1900W
thermo scientific™
AB1900W
White Case of 25 Each for $352.76
AB1900P
thermo scientific™
AB1900P
Purple Pack of 25 Each for $311.55
BC1900W
thermo scientific™
BC1900W
White Pack of 25 Each for $372.39
BC1900
thermo scientific™
BC1900
Clear Pack of 25 Each for $353.97
AB1900B
abgene inc usa
AB1900B
Blue Pack of 25 Each for $302.03
AB1900G
abgene inc usa
AB1900G
Green Pack of 25 Each for $302.03
AB1900R
abgene inc usa
AB1900R
Red Pack of 25 Each for $302.03
AB1900Y
abgene inc usa
AB1900Y
Yellow Pack of 25 Each for $311.55
Description & Specifications
Specifications
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
For Use With (Application) PCR, qPCR, RT-PCR
Volume (Metric) Well 100μL
Well Color Clear
Material Polypropylene
Rimmed Yes
Skirt Style Semi
The Thermo Scientific 96-Well Semi-Skirted, Fast Block, Barcoded Plates is compatible with all ABI Fast block machines, including Life Technologies PCR and qPCR thermal cyclers. These plates feature a raised deck to prevent spills and a cut corner at the A1 location.
The 96-Well Semi-Skirted, Fast Block, Barcoded Plates are available in white plates for qPCR applications and have clear wells for maximum sample visibility.
Highlights
  • Directly compatible with all Applied Biosystems thermal cycler fast blocks with no adapters necessary
  • Raised deck of plate to aid in spill prevention
  • 0.1 mL maximum well volume (when used with adhesive and heat seals)
  • Clear well for sample visibility
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut Corner: A1
Applications
  • PCR
  • qPCR/Real-Time PCR
The barcodes are the industry standard code 128. The label itself can withstand temperatures from -196°C up to 120°C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any 'rubbing off' of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.