Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ Thermo-Fast™ 96 PCR Detection Plate MKIII


96-well low-profile semi-skirted PCR plate for the fast block of Life Technologies PCR and qPCR thermal cyclers. Supplied with barcodes for sample and plate traceability.

Manufacturer: thermo scientific™  AB1900P

 View more versions of this product

Catalog No. AB1900P

  • / Pack of 25 Each
 

Description & Specifications

Specifications

For Use With (Application) PCR, qPCR, RT-PCR
Rimmed Yes
Skirt Style Semi
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
Well Color Clear
Color Purple
Material Polypropylene
Volume (Metric) Well 100μL
Product Type 96-Well PCR Plate
Applications PCR, qPCR, RT-PCR
Material Polypropylene