Please login to your online account to display your discounted pricing

ABgene™ Thermo-Fast™ 96 PCR Detection Plate MKIII


96-well low-profile semi-skirted PCR plate for the fast block of Life Technologies PCR and qPCR thermal cyclers. Supplied with barcodes for sample and plate traceability.

Manufacturer: abgene inc usa  AB1900G

 View more versions of this product

Catalog No. AB1900G

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Material Polypropylene
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
Well Color Clear
Product Type 96-Well PCR Plate
Applications PCR, qPCR, RT-PCR
Rimmed Yes
Skirt Style Semi
Volume (Metric) Well 100μL
For Use With (Application) PCR, qPCR, RT-PCR
Color Green
Material Polypropylene