Please login to your online account to display your discounted pricing

ABgene™ Thermo-Fast™ 96 PCR Detection Plate MKIII


96-well low-profile semi-skirted PCR plate for the fast block of Life Technologies PCR and qPCR thermal cyclers. Supplied with barcodes for sample and plate traceability.

Manufacturer: abgene inc usa  AB1900B

 View more versions of this product

Catalog No. AB1900B

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Volume (Metric) Well 100μL
Well Color Clear
Skirt Style Semi
Material Polypropylene
For Use With (Application) PCR, qPCR, RT-PCR
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
Color Blue
Rimmed Yes