Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ SuperPlate 96-Well PCR Plates


96-well semi-skirted PCR plates with a flat deck for improved sealing in PCR and qPCR applications.

Manufacturer: thermo scientific™  AB2800P

 View more versions of this product

Catalog No. AB2800P

  • / Case of 25 Each
 

Description & Specifications

Specifications

For Use With (Application) For improved sealing in PCR and qPCR applications
Material Polypropylene
Rimmed Yes
Skirt Style Full
Certifications/Compliance DNA, DNase, RNase-free
Volume (Metric) Well 200μL
Color Purple
Product Type SuperPlate 96-Well PCR Plate
Applications For improved sealing in PCR and qPCR applications
Material Polypropylene