Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ SuperPlate 96-Well PCR Plates


96-well semi-skirted PCR plates with a flat deck for improved sealing in PCR and qPCR applications.

Manufacturer: thermo scientific™  BC2800W

 View more versions of this product

Catalog No. BC2800W

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Material Polypropylene
Rimmed Yes
Certifications/Compliance DNA, DNase, RNase-free
For Use With (Application) For improved sealing in PCR and qPCR applications
Color White
Skirt Style Full
Volume (Metric) Well 200μL