Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ Plate Covers


Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking

Manufacturer: thermo scientific™  AB0752

 View more versions of this product

Catalog No. AB-0752

  • / Case of 50 Each
 

Description & Specifications

Specifications

No. per Case 50
With Stacking ridge
For Use With For all 96-well plates
Material Polystyrene
Format Plate Cover