Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ Plate Covers

Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking

$198.27 - $227.33

Specifications

Format Plate Cover
No. per Case 50
View More Specs
Products
Catalog Number Mfr. No. For Use With Material Price Quantity    

AB-0755

 
thermo scientific™
AB0755
For Thermo-fast 384-well plates Polypropylene Case of 50 Each for $201.67

AB-0751

 
thermo scientific™
AB0751
For all 96-well plates Polystyrene Case of 50 Each for $227.33

AB-0752

 
thermo scientific™
AB0752
For all 96-well plates Polystyrene Case of 50 Each for $198.27
Description & Specifications

Specifications

Format Plate Cover
No. per Case 50

Thermo Scientific Plate Covers provide effective sealing options for a variety of applications and can be used with 96-well plates. The covers are made of pollypropylene and are either ridged or non-ridged.

Highlights

  • Polystyrene covers can be used with all 96-well plates
  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates

Hazardous: No

Storage Condition: Ambient

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.