Please login to your online account to display your discounted pricing

Thermo Scientific™ 96-Well Semi-Skirted Plates, Raised Deck


96-Well Semi-Skirted Plates for PCR and qPCR applications.

Manufacturer: thermo scientific™  AB1100B

 View more versions of this product

Catalog No. AB-1100B

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Well Color Clear
For Use With (Application) PCR, qPCR applications
Certifications/Compliance DNA, DNase, RNase-free
Skirt Style Semi
Rimmed Yes
Color Blue
Material Polypropylene
Volume (Metric) Well 300μL
Accessories

Accessories for Catalog Number: AB1100B

Item Mfr. No. Item Description Price Per Unit Quantity  
AB-0427
Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats
thermo scientific™
AB0427
Round or square capped mats for sealing 96-well storage plates, available in autoclavable and piercable formats. Pack of 5 Each for $181.81