Please login to your online account to display your discounted pricing

Thermo Scientific™ 96-Well Semi-Skirted Plates, Raised Deck


96-Well Semi-Skirted Plates for PCR and qPCR applications.

Manufacturer: thermo scientific™  AB1100150

 View more versions of this product

Catalog No. AB1100150

  • / Case of 150 Each
 

Description & Specifications

Specifications

Certifications/Compliance DNA, DNase, RNase-free
Well Color Clear
Volume (Metric) Well 300μL
Color Natural
Material Polypropylene
Skirt Style Semi
For Use With (Application) PCR, qPCR applications
Rimmed Yes