Please login to your online account to display your discounted pricing

Thermo Scientific™ 96-Well Semi-Skirted Plates, Raised Deck


96-Well Semi-Skirted Plates for PCR and qPCR applications.

Manufacturer: thermo scientific™  BC1100

 View more versions of this product

Catalog No. BC1100

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Skirt Style Semi
Certifications/Compliance DNA, DNase, RNase-free
Well Color Clear
For Use With (Application) PCR, qPCR applications
Material Polypropylene
Rimmed Yes
Volume (Metric) Well 300μL
Color Clear