Please login to your online account to display your discounted pricing

Thermo Scientific™ 96-Well Semi-Skirted Plates, Raised Deck


96-Well Semi-Skirted Plates for PCR and qPCR applications.

Manufacturer: thermo scientific™  AB1100W

 View more versions of this product

Catalog No. AB-1100W

  • / Pack of 25 Each
 

Description & Specifications

Specifications

For Use With (Application) PCR, qPCR applications
Material Polypropylene
Rimmed Yes
Volume (Metric) Well 300μL
Well Color Clear
Color White
Skirt Style Semi
Certifications/Compliance DNA, DNase, RNase-free
Product Type 96-Well Microplate
Applications PCR, qPCR applications
Material Polypropylene
Accessories

Accessories for Catalog Number: AB1100W

Item Mfr. No. Item Description Price Per Unit Quantity