Please login to your online account to display your discounted pricing

Thermo Scientific™ 96-well Sealing Mats


Round or square capped mats for sealing 96-well storage plates, available in autoclavable and piercable formats.

Manufacturer: thermo scientific™  AB0662

 View more versions of this product

Catalog No. AB-0662

  • / Case of 50 Each
 

Description & Specifications

Specifications

Temperature Range (English) -4° to 131°F
Temperature Range (Metric) -20° to 55°C
Product Type Sealing Mat
Material TPE
Format Pierceable
For Use With 96-well storage plate
Applications Designed for sealing 96-well storage plate and suitable for long term storage
Under Well Shape Square
Autoclavable No
No. per Case 50