Thermo Scientific™ 96-Well Non-Skirted Plates, Low Profile
Low profile 96-well plates for use in PCR and qPCR applications.
$87.09 - $157.61
Specifications
Certifications/Compliance Certified free from human DNA, RNases, and DNases
For Use With (Application) PCR, qPCR applications
Volume (Metric) Well 200 μL
Material Polypropylene
Rimmed Yes
View More Specs
Products 8
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
AB-0700 View Documents Thermo Scientific™
AB0700
Clear Pack of 25 Each for $157.61
AB-0700G View Documents Thermo Scientific™
AB0700G
Green Pack of 25 Each for $157.61
AB-0700P View Documents Thermo Scientific™
AB0700P
Purple Pack of 25 Each for $157.61
AB-0700B View Documents Thermo Scientific™
AB0700B
Blue Pack of 25 Each for $157.61
AB-0700R View Documents Thermo Scientific™
AB0700R
Red Pack of 25 Each for $157.61
AB-0700Y View Documents Thermo Scientific™
AB0700Y
Yellow Pack of 25 Each for $157.61
AB-0700W View Documents Thermo Scientific™
AB0700W
White Pack of 25 Each for $87.09
AB0700L View Documents Thermo Scientific™
AB0700L
Clear w/Black Lettering Pack of 25 Each for $151.61
Description
Description
The low profile PCR plate has shorter wells than the standard profile plate (AB-0600), decreasing the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. These low profile products are especially recommended for use with reaction volumes below 20μL. They are available in assorted colors and are compatible with standard multichannel pipettes.
The low profile Thermo Scientific PCR plate has shorter wells than the standard profile plate (AB-0600), decreasing the “dead space” between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. These low profile products are especially recommended for use with reaction volumes below 20 μL. They are available in assorted colors and are compatible with standard multichannel pipettes.
Highlights
  • Low profile to reduce dead space and to increase PCR efficiency
  • Available with black alpha-numeric lettering
  • Improved access for liquid handling
  • Maximum fill volume of 0.2 mL
  • Cut corner: H12
Applications
  • PCR
  • qPCR/Real-Time PCR
Certifications
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Certified free from human DNA, RNases, and DNases
200 μL
Yes
PCR, qPCR applications
Polypropylene
Non-skirt