Please login to your online account to display your discounted pricing
Thermo Scientific™ 96-Well Non-Skirted Plates
96-well non-skirted plates for use in PCR and qPCR applications. Available with black lettering for improved sample tracking during pipetting.
$237.82 - $274.45
Products
Catalog Number Mfr. No. Price Quantity    
AB-0600
  Accessories
thermo scientific™
AB0600
Pack of 25 Each for $237.82
AB-0600B
  Accessories
thermo scientific™
AB0600B
Pack of 25 Each for $237.82
AB0600K
thermo scientific™
AB0600K
Case of 25 Each for $274.45
AB-0600W
  Accessories
thermo scientific™
AB0600W
Pack of 25 Each for $250.18
AB-0600Y
  Accessories
thermo scientific™
AB0600Y
Pack of 25 Each for $237.82
AB-0600R
  Accessories
thermo scientific™
AB0600R
Pack of 25 Each for $237.82
AB-0600P
  Accessories
thermo scientific™
AB0600P
Pack of 25 Each for $237.82
AB-0600G
  Accessories
thermo scientific™
AB0600G
Pack of 25 Each for $237.82
Description & Specifications
96-Well Non-Skirted Plates for use in PCR and qPCR applications hold up to 0.3 mL of well volume when paired with seals like adhesive films/foils, Thermo-Mat and cap strips. The plates feature a cut corner located at H1 and are compatible with most thermal cycles.
The well bottom is clear for maximum visibility. Available in assorted colors with clear or black alpha-numeric lettering. Compatible with standard multichannel pipettes.
The Thermo Scientific 96-Well Non-Skirted Plates for use in PCR and qPCR applications hold up to 0.3 mL of well volume when paired with seals like adhesive films/foils, Thermo-Mat and cap strips. The plates feature a cut corner located at H1 and are compatible with most thermal cycles.
The well bottom is clear for maximum visibility. Available in assorted colors with clear or black alpha-numeric lettering. Compatible with standard multichannel pipettes.
Highlights
  • Non-skirted format compatible with most thermal cyclers
  • Available with black alpha-numeric lettering
  • Cut corner: H1
  • Clear well bottom for sample visibility
  • 0.3 mL maximum well volume (when used with adhesive or heat seals)
  • Sealable using Thermo-Mat, Adhesive Films and Foils, Heat Seals and Flat and Domed Cap Strips
Applications
  • PCR
  • qPCR/Real-Time PCR
Certified free of human DNA, RNase, DNase, and endotoxins.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.