Thermo Scientific™ 8 x 15mL Swinging Bucket Rotor
Ideal for centrifugation of blood collection tubes and medical laboratories or practices with low sample throughput
$317.70 - $317.70
Products 1
Catalog Number Mfr. No. Capacity Type For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Capacity Type For Use With (Equipment) Price Quantity  
76-003-263 View Documents Thermo Scientific™
76003263
6 Tubes Bucket Rotor Thermo Scientific™ 8 x 15mL swing bucket rotor Each for $317.70
Description
Description
  • Economical small centrifuge with swinging bucket rotor
  • Ideal centrifugation of blood collection tubes
  • Complete system with 8-place swinging bucket rotor and buckets
  • Outstanding quality and reliability with induction drive and microprocessor control
  • Capacity: 8 x 15mL (0.12L)
  • Max. speed/RCF: 3600rpm/2028 x g
Order Info
Buckets (76003262) and pads (76003266) are sold separately; order rotor body (76003265) and components to complete assembly. Contact your Sales Representative for information.