missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ 8 x 15mL Swinging Bucket Rotor

Ideal for centrifugation of blood collection tubes and medical laboratories or practices with low sample throughput

Manufacturer:  Thermo Scientific™ 76003263

 View more versions of this product

Catalog No. 76-003-263



Description

Description

  • Economical small centrifuge with swinging bucket rotor
  • Ideal centrifugation of blood collection tubes
  • Complete system with 8-place swinging bucket rotor and buckets
  • Outstanding quality and reliability with induction drive and microprocessor control
  • Capacity: 8 x 15mL (0.12L)
  • Max. speed/RCF: 3600rpm/2028 x g

Order Info

Buckets (76003262) and pads (76003266) are sold separately; order rotor body (76003265) and components to complete assembly. Contact your Sales Representative for information.

Specifications

Specifications

Bucket Rotor
11.3cm
Thermo Scientific™ 8 x 15mL swing bucket rotor
6 Tubes
0.3cm
Documents